missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GILZ Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12352-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.Specifications
GILZ | |
Polyclonal | |
Unconjugated | |
TSC22D3 | |
DIP, DSIP immunoreactive peptide, DSIPI, GILZ, hDIP, Protein DIP, TSC 22 like protein, TSC 22 related protein, TSC 22R, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC22D3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14605, 1831, 83514 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99576, Q9EQZ1, Q9Z2S7 | |
TSC22D3 | |
GILZ Fusion Protein Ag3015 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |