Läs mer
Ginsenoside Rb2, MedChemExpress
Produktkod. 16451278
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16451278 5 mg
16448838 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16451278 Leverantör Medchem Express Leverantörsnummer HYN0040
MedChemExpress Ginsenoside Rb2 is one of the main bioactive components of ginseng extracts. Rb2 can upregulate GPR120 gene expression. Ginsenoside Rb2 has antiviral effects.
Ginsenoside Rb2 is one of the main bioactive components of ginseng extracts. Rb2 can upregulate GPR120 gene expression. Ginsenoside Rb2 has antiviral effects.
Specifikationer
Kemiskt namn eller material Ginsenoside Rb2
Fysisk form Solid
CAS 11021-13-9
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C53H90O22
För användning med (applikation) COVID-19-anti-virus
Synonym Ginsenoside C
Löslighetsinformation H2O : ≥ 100 mg/mL (92.66 mM) ∣DMSO : 9.5 mg/mL (8.80 mM; Need ultrasonic and warming)
LEDER C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O[C@@](O[C@H](CO)[C@@H](O)[C@@H]4O)([H])[C@@H]4O[C@]([C@@H]([C@@H](O)[C@@H]5O)O)([H])O[C@@H]5CO)C)(CC[C@@]2([H])C3(C)C)[C@]6([C@@]1([H])[C@]([C@@](CC/C=C(C)/C)(C)O[C@@H]([C@@H]([C@@H](O)[C@@H]7O)O)O[C@@H]7CO[C@H](OC[C@H](O)[C@@H]8O)[C@@H]8O)([H])CC6)C
Molekylvikt (g/mol) 1079.27
Formel vikt 1079.27
Procent renhet 98.0%