Läs mer
Ginsenoside Rf, MedChemExpress
Produktkod. 16491448
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16491448 5 mg 5mg
16409038 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16491448 Leverantör Medchem Express Leverantörsnummer HYN0601
MedChemExpress Ginsenoside Rf is a trace component of ginseng root. Ginsenoside Rf inhibits N-type Ca2+ channel.
Ginsenoside Rf is a trace component of ginseng root. Ginsenoside Rf inhibits N-type Ca2+ channel.
Specifikationer
Kemiskt namn eller material Ginsenoside Rf
CAS 52286-58-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C42H72O14
Kvantitet 5 mg
Synonym Panaxoside Rf
Löslighetsinformation DMSO : 100 mg/mL (124.84 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (62.42 mM; Need ultrasonic)
LEDER C[C@@]([C@@]12C)(C[C@H](O[C@@](O[C@H](CO)[C@@H](O)[C@@H]3O)([H])[C@@H]3O[C@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO)[C@@]5([H])C6(C)C)[C@@](C[C@@H](O)[C@]1([H])[C@]([C@@](C)(O)CC/C=C(C)/C)([H])CC2)([H])[C@]5(CC[C@@H]6O)C
Molekylvikt (g/mol) 801.01
Formel vikt 801.01
Procent renhet 95.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death