Läs mer
Ginsenoside Rh4, TRC
Produktkod. 30240843
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240843 5 mg
30240498 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG4102405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 174721-08-5
Molekylformel C36H60O8
Molekylvikt (g/mol) 620.86
Synonym (3beta,6alpha,12beta,20E)-3,12-Dihydroxydammara-20(22),24-dien-6-yl beta-D-glucopyranoside;
IUPAC-namn (2R,3R,4S,5S,6R)-2-[[(3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-3,12-dihydroxy-4,4,8,10,14-pentamethyl-17-[(2E)-6-methylhepta-2,5-dien-2-yl]-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-6-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
LEDER CC(=CC\C=C(/C)\[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O)C(C)(C)[C@@H]4[C@H](C[C@@]23C)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C
Specifikationer
Kemiskt namn eller material Ginsenoside Rh4
Kvantitet 5 mg
InChI formel InChI=1S/C36H60O8/c1-19(2)10-9-11-20(3)21-12-15-35(7)27(21)22(38)16-25-34(6)14-13-26(39)33(4,5)31(34)23(17-36(25,35)8)43-32-30(42)29(41)28(40)24(18-37)44-32/h10-11,21-32,37-42H,9,12-18H2,1-8H3/b20-11+/t21-,22-,23+,24-,25-,26+,27+,28-,29+,30-,31+,32-,34-,35-,36-/m1/s1
Formel vikt 620.4288