Läs mer
GIT1 Antibody (924640), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8508MFV610
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
GIT1 Monoclonal antibody specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
Specifikationer
| GIT1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, GPCR, Signal Transduction | |
| 28964 | |
| Store at 4C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF | |
| 924640 | |
| Western Blot, ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
| E. coli-derived recombinant human GIT1, Ser485-Asp636, Accession # Q9Y2X7 | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |