missing translation for 'onlineSavingsMsg'
Learn More

GJB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12880-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890824

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Gap junctions are conduits that allow the direct cell-to-cell passage of small cytoplasmic molecules, including ions, metabolic intermediates, and second messengers, and thereby mediate intercellular metabolic and electrical communication. Gap junction channels consist of connexin protein subunits, which are encoded by a multigene family. GJBs (gap-junction proteins or connexins) play crucial functional roles associated with these channels. Defects in GJB3 have been linked to erythrokeratodermia variabilis (EKV) is an autosomal domit genodermatosis characterized by transient figurate red patches or hyperkeratosis. Mutations in GJB2 have also been associated with genetically derived hearing impairments, including autosomal recessive nonsyndromic deafness.
Specifications

Specifications

GJB3
Polyclonal
Unconjugated
GJB3
Connexin 31, CX31, DFNA2, DFNA2B, EKV, Gap junction beta 3 protein, GJB3
Rabbit
Antigen Affinity Chromatography
RUO
14620, 2707, 29585
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75712, P25305, P28231
Gjb3
GJB3 Fusion Protein Ag3839
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers