Läs mer
Glabridin, TRC
Produktkod. 30005920
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005920 50 mg
17977994 10 mg
17958404 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG40640050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59870-68-7
Molekylformel C20 H20 O4
Molekylvikt (g/mol) 324.37
Synonym 1,3-Benzenediol, 4-[(3R)-3,4-dihydro-8,8-dimethyl-2H,8H-benzo[1,2-b:3,4-b']dipyran-3-yl]-, 1,3-Benzenediol, 4-(3,4-dihydro-8,8-dimethyl-2H,8H-benzo[1,2-b:3,4-b']dipyran-3-yl)-, (R)-, 2H,8H-Benzo[1,2-b:3,4-b']dipyran, 1,3-benzenediol deriv., 4-[(3R)-3,4-Dihydro-8,8-dimethyl-2H,8H-benzo[1,2-b:3,4-b']dipyran-3-yl]-1,3-benzenediol, Glabridin
IUPAC-namn 4-[(3R)-8,8-dimethyl-3,4-dihydro-2H-pyrano[2,3-f]chromen-3-yl]benzene-1,3-diol
LEDER CC1(C)Oc2ccc3C[C@@H](COc3c2C=C1)c4ccc(O)cc4O
Specifikationer
Kemiskt namn eller material Glabridin
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C20H20O4/c1-20(2)8-7-16-18(24-20)6-3-12-9-13(11-23-19(12)16)15-5-4-14(21)10-17(15)22/h3-8,10,13,21-22H,9,11H2,1-2H3/t13-/m0/s1
Formel vikt 324.1362
Procent renhet >95%