Läs mer
Glafenic Acid Hydrchloride Salt, TRC
Produktkod. 30403962
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30403962 100 mg
1 options
Produktkod. 30403962 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG406493100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Glafenic Acid Hydrchloride Salt
CAS 52803-56-2
Molekylformel C16H11ClN2O2 . HCl
Kvantitet 100 mg
Synonym 2-[(7-Chloro-4-quinolinyl)amino]-benzoic Acid Hydrochloride (1:1), 2-[(7-Chloro-4-quinolinyl)amino]-benzoic Acid Monohydrochloride (9CI)
InChI formel InChI=1S/C16H11ClN2O2.ClH/c17-10-5-6-11-14(7-8-18-15(11)9-10)19-13-4-2-1-3-12(13)16(20)21;/h1-9H,(H,18,19)(H,20,21);1H
LEDER Cl.OC(=O)c1ccccc1Nc2ccnc3cc(Cl)ccc23
IUPAC-namn 2-[(7-chloroquinolin-4-yl)amino]benzoic acid;hydrochloride
Molekylvikt (g/mol) 335.2
Formel vikt 334.0276