Learn More
Poulten Graf™ Glass BOD Bottle
Prevents air entrapment for your biological oxygen demand needs. Poulten Graf™ Glass BOD Bottle is made of soda lime Volac™ glass with an interchangeable stopper.
Brand: Poulten Graf™ GS748/A
Additional Details : Weight : 0.01000kg
Description
- Glass peg stopper adjusted to leave a "well" for water seal at bottle opening
- Pointed cone on stopper to prevent air entrapment
- Manufactured from clear glass
Specifications
Glass |