missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Glass Caps

Brand:  Duran™ 214412904

Additional Details : Weight : 0.43000kg

 View more versions of this product

Product Code. 15231749

  • 1837.98 SEK / Pack of 10


Estimated Shipment
28-12-2018
Add to basket

Specifications

Specifications

500mL
Bottle, Media Culture
46mm
10Pack
Glass Caps
Videos