Läs mer
Jaytec Glass™ Glass Dropping Pipets with Teat
Produktkod. 11366954
Change view
Examen:
1 mL
Förpackningsstorlek:
Paket med 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Examen Förpackningsstorlek
11366954 1 mL Paket med 12
11306804 - Paket med 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11366954 Leverantör Jaytec Glass™ Leverantörsnummer PMK380040X
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Includes natural rubber cap to assist in retrieving and dispensing liquid/media. Jaytec Glass™ Glass Dropping Pipets with Teat are available with our without graduations.
Natural rubber cap assists in retrieving and dispensing liquid/media.
Specifikationer
Material Glass, Rubber
Graderad Yes
Examen 1 mL
Inkluderar Teat
Kvantitet 12 Pack
Volym (metrisk) 2 mL
Typ Dropping Pipet