Learn More
Lenz Laborglasintrumente Glass Filter Plugs with Tube
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07208220
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11536784
  • 532.12 SEK / Each
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
  • Filter plugs
  • Cylindrical
  • With tube (dia. 8 mm x length 210 mm)
  • Material: borosilicate glass 3.3
Specifications
Specifications
Borosilicate Glass (3.3)
2
210 mm
9 mm