missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Glass Filter Plugs with Tube

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07208320

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11546784

  • 532.12 SEK / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

  • Filter plugs
  • Cylindrical
  • With tube (dia. 8 mm x length 210 mm)
  • Material: borosilicate glass 3.3
Specifications

Specifications

Borosilicate Glass (3.3)
3
210 mm
9 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers