Learn More
Hirschmann™ Glass Inserts
Exactly arranged to match with the most common patterns of pipetting robots. Hirschmann Instruments™ Glass Inserts provide high troughtput and low costs.
Brand: Hirschmann™ 9240596
Additional Details : Weight : 0.16200kg
Description
- Closed at bottom
- Chemically inert
- Lids made of silicone, PP or PTFE available
Specifications
For polypropylene Plates | |
Glass | |
96Pack | |
96/Pk. |