Learn More
Trajan Scientific and Medical™ Glass Lined Stainless Steel Union
Gives low dead volume connections for optimum chromatography.
1345.00 SEK
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
15229123
|
Trajan Scientific and Medical™
103420 |
0.5 mm |
1345.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
15239123
|
Trajan Scientific and Medical™
1034200 |
0.8 mm |
1345.00 SEK
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
Union | |
Stainless Steel |