Learn More
Trajan Scientific and Medical™ Glass Lined Stainless Steel Union
Gives low dead volume connections for optimum chromatography.
Brand: Trajan Scientific and Medical™ 103420
Additional Details : Weight : 0.01000kg
Description
- Low dead volume
- High temperature applications
- Leak free connections
Specifications
Union | |
Stainless Steel | |
0.5 mm |