missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Petri Dish, Glass

Shallow, lidded glass dishes for culturing cells including bacteria. Karl Hecht™ Petri Dishes, Glass are available in a range of convenient sizes.

Brand:  Karl Hecht™ 41042006

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10460552

  • 14.84 SEK / Each
Estimated Shipment: 19-04-2024
to see stock.
Add to basket

Description

Description

  • Glass construction.
  • Lidded.
  • Available in a variety of sizes.
Specifications

Specifications

Glass
Petri Dish
4 cm
Round
1.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers