Learn More
Hanna Instruments™ Glass pH Electrode
Brand: Hanna Instruments™ HI1217D
Additional Details : Weight : 0.50000kg
Specifications
DIN | |
pH Electrode | |
PEI | |
20°C to 40°C |
5 mm | |
General purpose pH electrode for HI 8314 and HI 9210N | |
0 to 13 |