Learn More
Carl Zeiss™ Glass Plate for TL-DF
Accessory DV4 clear glass plate replacement for TL-DF
Brand:  Carl Zeiss™ 475265-0001-000
Additional Details : Weight : 1.00000kg
Product Code. 11466797
  • 421.72 SEK / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
The DV4 has four illumination modes, which can be changed at the press of a button. The illumination modes are: brightfield reflected light; brightfield transmitted light; mixed light; darkfield transmitted light. A range of eyepiece adapters enables attachment of a digital camera; 35mm SLR camera or video camera.
  • A stereomicroscope ideal for universities, R&D companies, industry and equipment manufacturers
  • Robust, ergonomic design, ideal for long practical sessions
  • Easy to operate with complete flexibility: a wide range of stands, brackets, stages and illuminators to suit a variety of applications is available
  • The eyepiece micrometer, allows the user to measure at overview or magnified
Specifications
Specifications
Plate
Glass
1 Ea.