Learn More
DWK Life Sciences Glass Stopper, Clear, SBW Glass, Standard Taper Ground Joint, Octagonal flat head
Clear SBW glass ground stopper.
Brand:  DWK Life Sciences 246240604
Additional Details : Weight : 0.20000kg
Product Code. 15211989
  • 803.51 SEK / Pack of 10
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
Solid SBW glass stoppers can be used to close glassware items such as flasks or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head.
  • All glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to Standard Taper ground joint
  • Octagonal shaped head
Specifications
Specifications
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders
10/Pk.
SBW Glass
No