Learn More
DWK Life Sciences Kimble™ Ultra-Ware™ Glass Support Base
The Kimble™ Ultra-Ware™ Glass Support Base is suitable for use with Kimble™ Ultra-Ware™ Microfiltration Assemblies.
Brand: DWK Life Sciences 953726-0001
Additional Details : Weight : 0.80000kg
Description
- Product Type: Support Base
- Material: Glass
- Size: 25 mm
- For Use with: Kimble™ Ultra-Ware™ Microfiltration Assembly
Specifications
Not Autoclavable | |
Kimble Ultra-Ware Microfiltration Assembly | |
Non-sterile | |
25 mm |
Reusable | |
Glass | |
Support Base |