Learn More
Alfa Aesar™ Glassy Carbon Conical Crucible
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
Brand:  Alfa Aesar™ 039007.KT
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15483675
  • 4309.75 SEK / Each
Estimated Shipment date
from Supplier 26-04-2018
Add to basket
Specifications
Specifications
150mL
33mm
74mm
Glassy Carbon
Conical
64mm
Glassy Carbon Crucible