Learn More
Glassy carbon rod, 2mm (0.08in) dia, type 2, Thermo Scientific Chemicals
Widely used as an electrode material in electrochemistry, as well as for high temperature crucibles and as a component of some prosthetic devices. It is used in laboratories as well.
Brand: Thermo Scientific Alfa Aesar 037997.DM
Additional Details : CAS Number : 7440-44-0 Weight : 0.01000kg
Description
Glassy carbon rod, type 2 is widely used as an electrode material in electrochemistry, as well as for high temperature crucibles and as a component of some prosthetic devices. It is used in laboratories as well.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
MFCD00133992 | |
Insoluble in water. |
Tube | |
100 mm |
RUO – Research Use Only