Learn More
Glassy carbon rod, 6mm (0.24 in.) dia, type 2, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045005.DR
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glassy carbon rod | |
Tube | |
MFCD00133992 | |
200 mm |
RUO – Research Use Only