Läs mer
Glatiramer Acetate, TRC
Produktkod. 17990985
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17990985 100 mg -
17931475 10 mg -
17972475 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17990985 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG406800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147245-92-9
Molekylformel (C9H11NO3 • C6 H14N2O2 • C5H9NO4 • C3H7NO2)x • xC2H4O2
Molekylvikt (g/mol) 623.65
Synonym L-Alanine polymer with L-Glutamic Acid L-Lysine L-Tyrosine Acetate, L-Tyrosine polymer with L-Alanine L-Glutamic Acid L-Lysine Acetate, 511: PN: WO2010103292 PAGE: 64 claimed sequence, Cop 1, Copaxone, Copolymer 1, L-Glutamic Acid peptide with L-Alanine L-Lysine L-Tyrosine Acetate Salt, Protiramer, TV 5010;
IUPAC-namn L-Glutamic acid, polymer with L-alanine, L-lysine and L-tyrosine, acetate (salt)
LEDER N[C@H](C(O)=O)CCC(O)=O.N[C@H](C(O)=O)C.N[C@H](C(O)=O)CCCCN.CC(O)=O.N[C@H](C(O)=O)CC1=CC=C(O)C=C1
Specifikationer
Kemiskt namn eller material Glatiramer Acetate
Kvantitet 100 mg
InChI formel InChI=1S/C9H11NO3.C6H14N2O2.C5H9NO4.C3H7NO2.C2H4O2/c10-8(9(12)13)5-6-1-3-7(11)4-2-6;7-4-2-1-3-5(8)6(9)10;6-3(5(9)10)1-2-4(7)8;1-2(4)3(5)6;1-2(3)4/h1-4,8,11H,5,10H2,(H,12,13);5H,1-4,7-8H2,(H,9,10);3H,1-2,6H2,(H,7,8)(H,9,10);2H,4H2,1H3,(H,5,6);1H3,(H,3,4)/t8-;5-;3-;2-;/m0000./s1
Formel vikt 623.3014