Läs mer
Glipizide, TRC
Produktkod. 17964215
Change view
Kvantitet:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17964215 50 mg
17992685 500 mg
17964515 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17964215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG41022550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29094-61-9
Molekylformel C21 H27 N5 O4 S
Molekylvikt (g/mol) 445.54
Synonym Pyrazinecarboxamide, N-[2-[4-[[[(cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl- (9CI), Urea, 1-cyclohexyl-3-[[p-[2-(5-methylpyrazinecarboxamido)ethyl]phenyl]sulfonyl]- (8CI), N-[2-[4-[[[(Cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methyl-2-pyrazinecarboxamide, Aldiab, CP 28720, Digrin, Dipazide, Glibenese, Glibetin, Glican, Glidiab, Glipid, Glipizid, Glipizide, Gluco-Rite, Glucolip, Glucotrol, Glucotrol Xl, Glucozide, Glupitel, Glupizide, Glyde, Glydiazinamide, Glynase, K 4024, Melizide, Mindiab, Minidab, Minidiab, Minodiab, N-(2-(4-(N-(Cyclohexylcarbamoyl)sulfamoyl)phenyl)ethyl)-5-methylpyrazine-2-carboxamide, N-(4-[β-(5-Methylpyrazine-2-carboxamido)ethyl]benzenesulfonyl)-N'-cyclohexylurea, Napizide, Ozidia, Samarium(III) ionophore I, Semiglynase, Sucrazide, TK 1320
IUPAC-namn N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
LEDER Cc1cnc(cn1)C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3
Specifikationer
Kemiskt namn eller material Glipizide
Kvantitet 50 mg
InChI formel InChI=1S/C21H27N5O4S/c1-15-13-24-19(14-23-15)20(27)22-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,22,27)(H2,25,26,28)
Formel vikt 445.1784