Läs mer
Tocris Bioscience™ GLP-1 (7-37)
Endogenous bioactive GLP-1 receptor ligand
Brand: Tocris Bioscience™ 5374/1
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
GLP-1 (7-37) is a endogenous GLP-1 receptor ligand; bioactive and truncated form of GLP-1; insulinotropic hormone.Specifikationer
| 0.95 | |
| Store at -20°C | |
| Observed M.W.: 3355.71 | |
| C151H228N40O47 |
| 106612-94-6 | |
| Glucagon-Like Peptide 1 Receptor Agonists | |
| 1 mg |