missing translation for 'onlineSavingsMsg'
Läs mer

MedChemExpress GLP-2(1-33)(human)

Produktkod. 16492799
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
16492799 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492799 Supplier MedChemExpress Supplier No. HYP1024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GLP-2(1-33) (human) is an enteroendocrine hormone which can bind to the GLP-2 receptor and stimulate the growth of intestinal epithelium.

Specifications

Renhet 99.18%
CAS 223460-79-5
Innehåll och lagring Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
För användning med (applikation) Metabolism-sugar/lipid metabolism
Molekylvikt (g/mol) 3766.19 g/mol
Produkttyp Peptides
Kvantitet 1 mg
Frakt skick Room temperature
Löslighet H2O : 25 mg/mL (6.64 mM∣ultrasonic and adjust pH to 9 with NH4OH)
Synonym GLP-2 (human)∣ Glucagon-like peptide 2 (human)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.