missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GLPG 0974
GREENER_CHOICE

Product Code. 18790468
Change view
Click to view available options
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18790468 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18790468 Supplier Tocris Bioscience™ Supplier No. 5621/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent FFA2 antagonist

GLPG 0974 is a potent FFA2 antagonist (IC50 = 9 nM). Exhibits selectivity for FFA2 over FFA3. Inhibits acetate-induced human neutrophil migration in plasma or buffer. Also inhibits FFA2-induced expression of CD11b activation-specific epitope (AE) in human whole blood. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-[[[(2R)-1-(Benzo[b]thien-3-ylcarbonyl)-2-methyl-2-azetidinyl]carbonyl][(3-chlorophenyl)methyl]amino]butanoic acid
CAS 1391076-61-1
Kvantitet 10 mg
Mål Free Fatty Acid Receptor Antagonists
Molekylformel C25H25ClN2O4S
Formel vikt 485
Renhet >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.