missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLRX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566066
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: AAASGMESNTS SSLENLATAP VNQIQETISD NCVVIFSKTS CSYCTMA Highest antigen sequence identity to the following orthologs - mouse 73%, rat 73%.
GLRX2 Polyclonal antibody specifically detects GLRX2 in Human samples. It is validated for Immunocytochemistry.Specifications
GLRX2 | |
Polyclonal | |
Unconjugated | |
GLRX2 | |
1700010P22Rik; AI645710; bA101E13.1 (GRX2 glutaredoxin (thioltransferase) 2); CGI-133; GLRX2; glutaredoxin (thioltransferase) 2; glutaredoxin 2; glutaredoxin 2 (thioltransferase); glutaredoxin-2, mitochondrial; GRX2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NS18 | |
GLRX2 | |
Recombinant Human GLRX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only