Learn More
GLRX5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559985
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LVKKDKVVVF LKGTPEQPQC GFSNAVVQIL RLHGVRDYAA YNVLDDPELR QGIKDYSNWP TIPQVYLNGE FVGGCDILLQ MHQNGDLV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
GLRX5 is monothiol glutaredoxin involved in the biogenesis of iron-sulfur clusters. GLRX5 required for normal iron homeostasis. GLRX5 is required for normal regulation of hemoglobin synthesis by the iron-sulfur protein ACO1.Specifications
GLRX5 | |
Polyclonal | |
Unconjugated | |
GLRX5 | |
2310004O13Rik; 2900070E19Rik; AU020725; C14orf87; FLB4739; GLRX5; glutaredoxin 5; glutaredoxin 5 homolog; glutaredoxin 5 homolog (S. cerevisiae); glutaredoxin-related protein 5, mitochondrial; GRX5; Monothiol glutaredoxin-5; PR01238; PRO1238; PRSA; RGD1308383; SIDBA3; SPAHGC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SX6 | |
GLRX5 | |
Recombinant protein corresponding to Human GLRX5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only