missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLS Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3A12A1, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | GLS |
---|---|
Clone | 3A12A1 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827985
|
Proteintech
66265-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817985
|
Proteintech
66265-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes the K-type mitochondrial glutaminase. The encoded protein is an phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. This protein is primarily expressed in the brain and kidney plays an essential role in generating energy for metabolism, synthesizing the brain neurotransmitter glutamate and maintaining acid-base balance in the kidney. Alternate splicing results in multiple transcript variants.Specifications
GLS | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AAD20, GAC, GLS, GLS1, glutaminase, K glutaminase, KIAA0838, L glutamine amidohydrolase | |
GLS | |
IgG2b | |
Protein A | |
Antibody |
3A12A1 | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Porcine, Mouse, Human, Rat | |
D3Z7P3, O94925, P13264 | |
14660, 24398, 2744, 399525 | |
GLS Fusion Protein Ag24227 | |
Primary | |
-20°C | |
GLS |