Learn More
GLT8D1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552711
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VPNALRHAVD GRQEEIPVVI AASEDRLGGA IAAINSIQHN TRSNVIFYIV TLNNTADHLR SWLNSDSLKS IRYKIVNFDP KLLEGKVKED PDQGESMKPL TFARFYLPIL VPSAKKAIYM DDDVIVQG Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
GLT8D1 Polyclonal antibody specifically detects GLT8D1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
GLT8D1 | |
Polyclonal | |
Unconjugated | |
Glt8d1 | |
2410004H05Rik; 5430414N14Rik; AD-017; AI450005; Da2-24; GALA4A; Glt8d1; glycosyltransferase 8 domain containing 1; glycosyltransferase 8 domain-containing protein 1; glycosyltransferase AD-017; MSTP137; MSTP139; UNQ572/PRO1134 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55830 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68CQ7 | |
Glt8d1 | |
Recombinant protein corresponding to Human GLT8D1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only