missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLTP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10850-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Accelerates the intermembrane transfer of various glycolipids. Catalyzes the transfer of various glycosphingolipids between membranes but does not catalyze the transfer of phospholipids. May be involved in the intracellular translocation of glucosylceramides.Specifications
GLTP | |
Polyclonal | |
Unconjugated | |
GLTP | |
GLTP, glycolipid transfer protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288707, 51228, 56356 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B0BNM9, Q9JL62, Q9NZD2 | |
Gltp | |
GLTP Fusion Protein Ag1293 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |