Läs mer
Glubionate Calcium Hydrate, TRC
Produktkod. 30405460
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30405460 1 g
1 options
Produktkod. 30405460 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG4127501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Calcium Glubionate
CAS 12569-38-9
Molekylformel C12 H21 O12 . C6 H11 O7 . Ca . H2 O
Kvantitet 1 g
Synonym Calcium, (4-O-β-D-galactopyranosyl-D-gluconato)(D-gluconato)-, monohydrate, D-Gluconic acid, 4-O-β-D-galactopyranosyl-, calcium complex, D-Gluconic acid, calcium complex, Calcium glubionate, Calcium glucono-galacto-gluconate, Calcium gluconolactobionate
InChI formel InChI=1S/C12H22O12.C6H12O7.Ca.H2O/c13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12;7-1-2(8)3(9)4(10)5(11)6(12)13;;/h3-10,12-20H,1-2H2,(H,21,22);2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t3-,4-,5+,6+,7-,8-,9-,10-,12+;2-,3-,4+,5-;;/m11../s1
LEDER O.[Ca+2].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-].OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(=O)[O-]
IUPAC-namn calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;(2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanoate;hydrate
Molekylvikt (g/mol) 610.529
Formel vikt 610.127