Läs mer
Glucose-6-phosphate dehydrogenase, from Leuconostoc mesenteroides, ammonium sulfate suspension, Thermo Scientific™
Produktkod. 15865548 Handla allt Thermo Scientific Produkter
Change view
Kvantitet:
1 kU
10 kU
100 kU
5 kU
Förpackningsstorlek:
100ku
10ku
1kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15865548 1 kU 1kg
15875548 5 kU 5kg
15835148 10 kU 10ku
15825148 100 kU 100ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 15865548 Leverantör Thermo Scientific Leverantörsnummer J16171.4I
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 9001-40-5
MDL-nummer MFCD00081656
Synonym E.C.1.1.1.49
Specifikationer
Kemiskt namn eller material Glucose-6-phosphate dehydrogenase
Kvantitet 1 kU
Fysisk form Suspension