Läs mer
Glufosinate, TRC
Produktkod. 17947279
Change view
Kvantitet:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17947279 2 mg
17978409 10 mg
17963855 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17947279 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG5969582MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51276-47-2
Molekylformel C5 H12 N O4 P
Molekylvikt (g/mol) 181.13
Synonym 2-Amino-4-(hydroxymethylphosphinyl)butanoic acid, (±)-Phosphinothricin, 3-Amino-3-carboxypropylmethylphosphinic acid, DL-2-Amino-4-(methylphosphino)butanoic acid, DL-Phosphinothricin, Glufosinate, HOE 35956
IUPAC-namn 2-amino-4-[hydroxy(methyl)phosphoryl]butanoic acid
LEDER CP(=O)(O)CCC(N)C(=O)O
Specifikationer
Kemiskt namn eller material Glufosinate
Anteckningar om renhetsgrad HPLC
Kvantitet 2 mg
InChI formel InChI=1S/C5H12NO4P/c1-11(9,10)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)(H,9,10)
Formel vikt 181.0504
Procent renhet >95%