missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20436-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody detected 60-70 kDa fully glycosylated, matured GLUT2 and a nonglycosylated form of GLUT2 around 38-45 kDa.
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.Specifications
GLUT2 | |
Polyclonal | |
Unconjugated | |
SLC2A2 | |
GLUT 2, GLUT2, SLC2A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20526, 25351, 6514 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P11168, P12336, P14246 | |
Slc2a2 | |
GLUT2 Fusion Protein Ag14204 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |