Läs mer
Glutaconic Acid, TRC
Produktkod. 30136699
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136699 10 g
30136047 5 g
30006620 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG59694510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1724-02-3
Molekylformel C5H6O4
Molekylvikt (g/mol) 130.1
Synonym 1,3-Propenedicarboxylic Acid, 1-Propene-1,3-dicarboxylic Acid, Pentenedioic Acid, 2-Pentenedioic Acid
LEDER OC(=O)C\C=C\C(=O)O
Specifikationer
Kemiskt namn eller material Glutaconic Acid
Kvantitet 10 g
InChI formel InChI=1S/C5H6O4/c6-4(7)2-1-3-5(8)9/h1-2H,3H2,(H,6,7)(H,8,9)/b2-1+