Learn More
Glutaminase C (GAC) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540135
Additional Details : Weight : 0.00910kg
Description
This antibody specifically recognizes GAC but not the KGA isoform. Predicted reactivity: Bovine (100%). Recommended positive controls: MDA-MB-231 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Glutaminase C (GAC) Polyclonal antibody specifically detects Glutaminase C (GAC) in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Glutaminase C (GAC) | |
Polyclonal | |
Unconjugated | |
GLS | |
6330442B14; AAD20; AI314027; B230365M23Rik; GAC; GAM; GLS; Gls1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; RATGLUT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2744 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O94925 | |
GLS | |
Recombinant protein encompassing a sequence within the C-terminus region of human Glutaminase C. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |