missing translation for 'onlineSavingsMsg'
Läs mer

Glutamine Synthetase Antibody (CL3004) [DyLight 405], R&D Systems™

Produktkod. 30290461 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
Förpackningsstorlek:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
30290461 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30290461 Leverantör Novus Biologicals Leverantörsnummer NBP246644V
 Begränsad artikel
Beräknad leverans: 02-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Användningsområden Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Klassificering Monoclonal
Klona CL3004
Konjugera DyLight 405
Formulering 50mM Sodium Borate
Gene Alias cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Värd art Mouse
Immunogen This antibody was developed using a recombinant protein derived from P15104, with the exact immunogen sequence remaining proprietary.
Reningsmetod Protein A purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Hypoxia, Neurodegeneration, Neuroscience, Vision
Primär eller sekundär Primary
Gen-ID (Entrez) 2752
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4⪚C in the dark.
Produkttyp Antibody
Form Purified
Isotyp IgG1
Produktlinje Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.