missing translation for 'onlineSavingsMsg'
Learn More

Glutamine synthetase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11037-2-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877833

  • 3248.87 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Structural maintece of chromosomes (SMC) and non-SMC condensin proteins associate into complexes that have been implicated in the process of chromosome condensation. A crucial prerequisite for accurate segregation of replicated sister chromatids is the condensation of the chromosomes into a manageable form prior to metaphase. The condensin I complex consists of two SMC subunits, SMC2 and SMC4, and three non-SMC subunits, CAP-H, CAP-G, and CAP-D2. An alternative complex, the condensin II complex, contains alternate non-SMC subunits, CAP-G2, CAP-H2, and CAP-D3. CAP-H2 is also known as Non-SMC condensin II complex, subunit H2 (NCAPH2) or kleisin beta isoform 2. The three non-SMC subunits in the condensing complexes form a regulatory subcomplex that is required to activate the SMC ATPases and to promote mitosis-specific chromatin binding of the holocomplex. The precise individual functions of each non-SMC protein in activation remain to be determined.
Specifications

Specifications

Glutamine synthetase
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09606, P15104, P15105
GLNS, GLUL, Glutamate ammonia ligase, Glutamate decarboxylase, Glutamine synthetase, GS, PIG43, PIG59
Rabbit
IgG
150 μL
-20°C
Primary
14645, 24957, 2752
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GLUL
Liquid
GLUL
Glutamine synthetase Fusion Protein Ag1510
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers