Läs mer
Glutaminyl Cyclase Inhibitor 1, MedChemExpress
Produktkod. 16464608
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16464608 10 mg
16460907 50 mg
16435456 100 mg
16417327 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16464608 Leverantör Medchem Express Leverantörsnummer HY112269
MedChemExpress Glutaminyl Cyclase Inhibitor 1 is a glutaminyl cyclase inhibitor with an IC50 of 0.5 μM.
Glutaminyl Cyclase Inhibitor 1 is a glutaminyl cyclase inhibitor with an IC50 of 0.5 μM.
Specifikationer
Kemiskt namn eller material Glutaminyl Cyclase Inhibitor 1
CAS 2110449-60-8
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C21H24FN3O2
Kvantitet 10 mg
Löslighetsinformation Ethanol : 100 mg/mL (270.69 mM; Need ultrasonic) ∣DMSO : 83.33 mg/mL (225.56 mM; Need ultrasonic)
LEDER FC1=CC=CC(C2=CC(OC)=C(OC)C=C2)=C1NCCCN3C=C(C)N=C3
Molekylvikt (g/mol) 369.43
Formel vikt 369.43
Procent renhet 98.34%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neurodegeneration