Läs mer
Glutaraldehydonitrile Diethyl Acetal, TRC
Produktkod. 30406403
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30406403 250 mg
1 options
Produktkod. 30406403 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG597310250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Glutaraldehydonitrile Diethyl Acetal
CAS 1468-47-9
Molekylformel C9H17NO2
Kvantitet 250 mg
Synonym 5,5-Diethoxy-pentanenitrile, 4-Cyano-butyraldehyde Diethyl Acetal, 5,5-Diethoxypentanenitrile, 5,5-Diethoxy-valeronitrile
InChI formel InChI=1S/C9H17NO2/c1-3-11-9(12-4-2)7-5-6-8-10/h9H,3-7H2,1-2H3
LEDER CCOC(CCCC#N)OCC
IUPAC-namn 5,5-diethoxypentanenitrile
Molekylvikt (g/mol) 171.24
Formel vikt 171.1259