Läs mer
Glycerol 1-Monobutyrate (Technical Grade), TRC
Produktkod. 17984625
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17984625 250 mg
17981945 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17984625 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG598415250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 557-25-5
Molekylformel C7H14O4
Molekylvikt (g/mol) 162.18
Synonym 1-Monobutyrin, 2,3-Dihydroxypropyl Butyrate, Glycerol 1-Monobutyrate, Glycerol α-Monobutyrate, Glycerol-α-mono-n-butyrate, Glyceryl 3-Monobutyrate NSC 8451, α-Monobutyrin
IUPAC-namn 2,3-dihydroxypropyl butanoate
LEDER CCCC(=O)OCC(O)CO
Specifikationer
Kemiskt namn eller material Glycerol 1-Monobutyrate
Kvantitet 250 mg
InChI formel InChI=1S/C7H14O4/c1-2-3-7(10)11-5-6(9)4-8/h6,8-9H,2-5H2,1H3
Formel vikt 162.0892