Läs mer
Glycerol 1-Monobutyrate (Technical Grade), TRC
Produktkod. 30407904
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30407904 1 g
1 options
Produktkod. 30407904 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG5984151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Glycerol 1-Monobutyrate
CAS 557-25-5
Molekylformel C7H14O4
Kvantitet 1 g
Synonym 1-Monobutyrin, 2,3-Dihydroxypropyl Butyrate, Glycerol 1-Monobutyrate, Glycerol α-Monobutyrate, Glycerol-α-mono-n-butyrate, Glyceryl 3-Monobutyrate NSC 8451, α-Monobutyrin
InChI formel InChI=1S/C7H14O4/c1-2-3-7(10)11-5-6(9)4-8/h6,8-9H,2-5H2,1H3
LEDER CCCC(=O)OCC(O)CO
IUPAC-namn 2,3-dihydroxypropyl butanoate
Molekylvikt (g/mol) 162.18
Formel vikt 162.0892