Läs mer
Glycerol Kinase Antibody, Allophycocyanin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-78018APC
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Glycerol Kinase Polyclonal antibody specifically detects Glycerol Kinase in Bacteria samples. It is validated for Western Blot, ELISA
Specifikationer
| Glycerol Kinase | |
| Polyclonal | |
| PBS | |
| Goat | |
| Multi-step | |
| RUO | |
| Primary | |
| Bacteria | |
| Purified |
| Western Blot, ELISA | |
| APC | |
| ATP:glycerol 3-phosphotransferase, EC 2.7.1.30, GK1, GKD, Glycerokinase, glycerol kinase | |
| Glycerol Kinase [Cellulomonas species] (Uniprot: D0VZG4) | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 2710 | |
| Store at 4°C in the dark. | |
| IgG |