Läs mer
Glycidamide- 13C3, TRC
Produktkod. 30136260
Change view
Kvantitet:
0.5 mg
2.5 mg
Förpackningsstorlek:
0.5mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136260 2.5 mg 2.5mg
18001430 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136260 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG6152532.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1216449-31-8
Molekylformel 13C3 H5 N O2
Molekylvikt (g/mol) 90.06
IUPAC-namn (2,3-^{13}C_{2})oxirane-2-carboxamide
LEDER N[13C](=O)[13CH]1[13CH2]O1
Specifikationer
Kemiskt namn eller material Glycidamide 13C3
Kvantitet 2.5 mg
InChI formel InChI=1S/C3H5NO2/c4-3(5)2-1-6-2/h2H,1H2,(H2,4,5)/i1+1,2+1,3+1
Formel vikt 90.0421