Läs mer
Glycidyl Caprate, TRC
Produktkod. 30240864
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240864 100 mg
30240841 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240864 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG615500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 26411-50-7
Molekylformel C13H24O3
Molekylvikt (g/mol) 228.33
Synonym Decanoic Acid 2-Oxiranylmethyl Ester, Decanoic Acid 2,3-Epoxypropyl Ester, 2,3-Epoxy-1-propanol Decanoate, Glycidyl Decanoate;
IUPAC-namn oxiran-2-ylmethyl decanoate
LEDER CCCCCCCCCC(=O)OCC1CO1
Specifikationer
Kemiskt namn eller material Glycidyl Caprate
Kvantitet 100 mg
InChI formel InChI=1S/C13H24O3/c1-2-3-4-5-6-7-8-9-13(14)16-11-12-10-15-12/h12H,2-11H2,1H3
Formel vikt 228.1725