Läs mer
Glycidyl Hexanoate, TRC
Produktkod. 30240639
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240639 100 mg
30240546 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240639 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG615560100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17526-74-8
Molekylformel C9H16O3
Molekylvikt (g/mol) 172.22
Synonym Hexanoic Acid 2-Oxiranylmethyl Ester, Hexanoic Acid 2,3-Epoxypropyl Ester, Hexanoic Acid Oxiranylmethyl Ester;
LEDER CCCCCC(=O)OCC1CO1
Specifikationer
Kemiskt namn eller material Glycidyl Hexanoate